Learn More
Medchemexpress LLC Zaldaride maleate | 109826-27-9 | MFCD04113002 | 99.7% | 544.60 | C30H32N4O6 | 1 MG
SDP
Catalog No. 5000244275
Encompass_Preferred
Zaldaride maleate is a potent, orally active, and selective calmodulin inhibitor used in biochemical and cellular research. It inhibits calmodulin-stimulated cAMP phosphodiesterase activity (IC50 ≈ 3.3 nM), modulates ion-channel currents, and is supplied for laboratory research use only.
  • Potent calmodulin inhibitor.
  • Orally active in pharmacological studies.
  • Low nanomolar IC50 for cAMP phosphodiesterase inhibition.
  • Reversibly blocks voltage-activated Na+, Ca2+, and K+ currents.
  • High purity suitable for research applications.
  • Provided as a small solid quantity for laboratory use.
Catalog No. 50-002-44275 Supplier Medchemexpress LLC Supplier No. HY105118A1MG
Zaldaride maleate is a potent, orally active, and selective calmodulin inhibitor used in biochemical and cellular research. It inhibits calmodulin-stimulated cAMP phosphodiesterase activity (IC50 ≈ 3.3 nM), modulates ion-channel currents, and is supplied for laboratory research use only.
  • Potent calmodulin inhibitor.
  • Orally active in pharmacological studies.
  • Low nanomolar IC50 for cAMP phosphodiesterase inhibition.
  • Reversibly blocks voltage-activated Na+, Ca2+, and K+ currents.
  • High purity suitable for research applications.
  • Provided as a small solid quantity for laboratory use.