Learn More
Medchemexpress LLC Zaldaride maleate | 109826-27-9 | MFCD04113002 | 99.7% | 544.60 g/mol | C30H32N4O6 | 10 MG
SDP
Catalog No. 5000243556
Encompass_Preferred
Zaldaride maleate is a research-grade calmodulin inhibitor used in biochemical and pharmacological studies to block calmodulin-mediated signaling. It is described as a potent, orally active, and selective antagonist of calmodulin and is applied in cellular assays and mechanistic investigations involving CaM-dependent pathways.
  • Potent calmodulin inhibition for mechanistic studies.
  • High purity suitable for research applications.
  • Supplied in small-mass formats for dose-ranging experiments.
  • Solid storage recommended at 4°C; solutions stable at -20°C to -80°C per guidance.
  • Molecular weight approximately 544.6 g/mol for dosing calculations.
Catalog No. 50-002-43556 Supplier Medchemexpress LLC Supplier No. HY105118A10MG
Zaldaride maleate is a research-grade calmodulin inhibitor used in biochemical and pharmacological studies to block calmodulin-mediated signaling. It is described as a potent, orally active, and selective antagonist of calmodulin and is applied in cellular assays and mechanistic investigations involving CaM-dependent pathways.
  • Potent calmodulin inhibition for mechanistic studies.
  • High purity suitable for research applications.
  • Supplied in small-mass formats for dose-ranging experiments.
  • Solid storage recommended at 4°C; solutions stable at -20°C to -80°C per guidance.
  • Molecular weight approximately 544.6 g/mol for dosing calculations.