Learn More
Medchemexpress LLC HY-B0619 10mg Medchemexpress, Zaltoprofen CAS:74711-43-6 Purity:>98%
SDP
Catalog No. 502082622
Encompass_Preferred
Medchemexpress, HY-B0619 10mg Zaltoprofen CAS:74711-43-6 Zaltoprofen (CN100), a non-steroidal anti-inflammatory drug (NSAID), is a preferential and orally active COX-2 inhibitor, with IC50s of 1.3 and 0.34 μM for COX-1 and COX-2, respectively. Zaltoprofen exhibits powerful anti-inflammatory effects as well as an analgesic action on inflammatory pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2622 Supplier Medchemexpress LLC Supplier No. HYB061910MG
Medchemexpress, HY-B0619 10mg Zaltoprofen CAS:74711-43-6 Zaltoprofen (CN100), a non-steroidal anti-inflammatory drug (NSAID), is a preferential and orally active COX-2 inhibitor, with IC50s of 1.3 and 0.34 μM for COX-1 and COX-2, respectively. Zaltoprofen exhibits powerful anti-inflammatory effects as well as an analgesic action on inflammatory pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.