Learn More
Cayman Chemical Zaltoprofen, Purity: ≥98%, CAS: 74711-43-6, 10 mg
SDP
Catalog No. 502969301
Encompass_Preferred
An NSAID that displays slight preferential inhibition for COX-2 (IC50s = 1.3 and 0.34 μM for COX-1 and COX-2, respectively); also inhibits bradykinin-induced nociceptive responses by blocking the activation of PKC
Catalog No. 50-296-9301 Supplier Cayman Chemical Supplier No. 1466210 MG
An NSAID that displays slight preferential inhibition for COX-2 (IC50s = 1.3 and 0.34 μM for COX-1 and COX-2, respectively); also inhibits bradykinin-induced nociceptive responses by blocking the activation of PKC