Learn More
Apexbio Technology LLC Zaltoprofen 74711-43-6 10mg
SDP
Catalog No. 502240781
Encompass_Preferred
Zaltoprofen (CAS 74711-43-6) is a nonsteroidal anti-inflammatory small molecule that functions as a cyclooxygenase (COX) inhibitor with a moderate selectivity for COX-2 over COX-1 (IC50 values of 0 34 M for COX-2 and 1 3 M for COX-1) In addition to inhibiting prostaglandin synthesis through COX modulation zaltoprofen has been shown to suppress bradykinin-induced nociceptive responses independent of its COX-inhibitory activity This multifaceted mechanism makes zaltoprofen a relevant tool for investigating inflammatory pathways and pain signaling in biomedical research
This item is not returnable. View return policy
Catalog No. 50-224-0781 Supplier Apexbio Technology LLC Supplier No. B146010
Zaltoprofen (CAS 74711-43-6) is a nonsteroidal anti-inflammatory small molecule that functions as a cyclooxygenase (COX) inhibitor with a moderate selectivity for COX-2 over COX-1 (IC50 values of 0 34 M for COX-2 and 1 3 M for COX-1) In addition to inhibiting prostaglandin synthesis through COX modulation zaltoprofen has been shown to suppress bradykinin-induced nociceptive responses independent of its COX-inhibitory activity This multifaceted mechanism makes zaltoprofen a relevant tool for investigating inflammatory pathways and pain signaling in biomedical research