Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zaltoprofen 98.0+%, TCI America™
SDP

Catalog No. Z00211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00211G 1 g
Z00215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z00211G Supplier TCI America Supplier No. Z00211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74711-43-6
Molecular Formula C17H14O3S
Molecular Weight (g/mol) 298.356
MDL Number MFCD00864323
InChI Key MUXFZBHBYYYLTH-UHFFFAOYSA-N
Synonym 2-(10,11-Dihydro-10-oxodibenzo[b,f]thiepin-2-yl)propionic Acid
PubChem CID 5720
IUPAC Name 2-(6-oxo-5H-benzo[b][1]benzothiepin-3-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)SC3=CC=CC=C3C(=O)C2)C(=O)O

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 1 g
Formula Weight 298.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Zaltoprofen
RTECSNumber HQ2526700
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.