Learn More
Medchemexpress LLC Zalunfiban dihydrochloride | 2815778-41-5 | 99.1% | 1 ML
SDP
Catalog No. 5000233073
Encompass_Preferred
Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist (reported IC50 ≈ 45 nM) provided as the dihydrochloride salt for research applications, including studies of platelet aggregation and myocardial infarction.
  • Potent αIIbβ3 antagonist with reported IC50 ≈ 45 nM.
  • Provided as the dihydrochloride salt, soluble in DMSO.
  • High purity (99.07%).
  • Available as a 10 mM solution in 1 mL vials and in multiple solid sizes.
  • Intended for preclinical and biochemical research use.
Catalog No. 50-002-33073 Supplier Medchemexpress LLC Supplier No. HY119350B10MM 1ML
Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist (reported IC50 ≈ 45 nM) provided as the dihydrochloride salt for research applications, including studies of platelet aggregation and myocardial infarction.
  • Potent αIIbβ3 antagonist with reported IC50 ≈ 45 nM.
  • Provided as the dihydrochloride salt, soluble in DMSO.
  • High purity (99.07%).
  • Available as a 10 mM solution in 1 mL vials and in multiple solid sizes.
  • Intended for preclinical and biochemical research use.