Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zalunfiban dihydrochloride | 2815778-41-5 | 99.1% | 10 MG
SDP

Catalog No. 5000233595
Encompass_Preferred

Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist (IC50 45 nM) supplied as a high-purity solid for in vitro and in vivo research on platelet function and myocardial infarction.

  • CAS number: 2815778-41-5
  • Molecular formula: C16H20Cl2N8O2S
  • Molecular weight: 459.35 g/mol
  • Purity: 99.1%
  • Appearance: Solid
  • In vitro solubility: DMSO 10 mg/mL (21.77 mM) with sonication and warming
  • Suggested storage: 4°C sealed, away from moisture; in solvent -80°C (6 months)
  • Typical pack size: 10 mg

Catalog No. 50-002-33595 Supplier Medchemexpress LLC Supplier No. HY119350B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist (IC50 45 nM) supplied as a high-purity solid for in vitro and in vivo research on platelet function and myocardial infarction.

  • CAS number: 2815778-41-5
  • Molecular formula: C16H20Cl2N8O2S
  • Molecular weight: 459.35 g/mol
  • Purity: 99.1%
  • Appearance: Solid
  • In vitro solubility: DMSO 10 mg/mL (21.77 mM) with sonication and warming
  • Suggested storage: 4°C sealed, away from moisture; in solvent -80°C (6 months)
  • Typical pack size: 10 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.