Learn More
Medchemexpress LLC Zalunfiban dihydrochloride | 2815778-41-5 | 99.1% | 100 MG
SDP
Catalog No. 5000234238
Encompass_Preferred
Zalunfiban dihydrochloride is the dihydrochloride salt of RUC-4, a potent, selective platelet αIIbβ3 antagonist (IC50 = 45 nM) used in preclinical and in vitro research of platelet inhibition and myocardial infarction.
  • Potent, selective αIIbβ3 antagonist (IC50 = 45 nM).
  • Suitable for platelet function and myocardial infarction research.
  • High purity (99.1%) as supplied.
  • Soluble in DMSO (10 mg/mL) with warming and sonication.
  • Store sealed, away from moisture; follow recommended cold storage for solutions.
Catalog No. 50-002-34238 Supplier Medchemexpress LLC Supplier No. HY119350B100MG
Zalunfiban dihydrochloride is the dihydrochloride salt of RUC-4, a potent, selective platelet αIIbβ3 antagonist (IC50 = 45 nM) used in preclinical and in vitro research of platelet inhibition and myocardial infarction.
  • Potent, selective αIIbβ3 antagonist (IC50 = 45 nM).
  • Suitable for platelet function and myocardial infarction research.
  • High purity (99.1%) as supplied.
  • Soluble in DMSO (10 mg/mL) with warming and sonication.
  • Store sealed, away from moisture; follow recommended cold storage for solutions.