Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zalunfiban dihydrochloride | 2815778-41-5 | 99.1% | 25 MG
SDP

Catalog No. 5000233202
Encompass_Preferred

Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist used in research of platelet aggregation and myocardial infarction. Supplied as the dihydrochloride salt, it is intended for in vitro and in vivo experimental studies.

  • Potent αIIbβ3 antagonist with IC50 = 45 nM.
  • High purity for research use (99.1%).
  • Chemical formula C16H20Cl2N8O2S and molecular weight 459.35 g/mol.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Suitable for studies of myocardial infarction and platelet aggregation.

Catalog No. 50-002-33202 Supplier Medchemexpress LLC Supplier No. HY119350B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist used in research of platelet aggregation and myocardial infarction. Supplied as the dihydrochloride salt, it is intended for in vitro and in vivo experimental studies.

  • Potent αIIbβ3 antagonist with IC50 = 45 nM.
  • High purity for research use (99.1%).
  • Chemical formula C16H20Cl2N8O2S and molecular weight 459.35 g/mol.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Suitable for studies of myocardial infarction and platelet aggregation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.