Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zalunfiban dihydrochloride | 2815778-41-5 | 99.1% | 5 MG
SDP

Catalog No. 5000234329
Encompass_Preferred

Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist used in research applications such as myocardial infarction studies. It is supplied as the dihydrochloride salt with high reported purity and is typically used in biochemical and pharmacological assays.

  • Potent αIIbβ3 antagonist (IC50 = 45 nM).
  • Reported purity 99.07%.
  • Molecular formula C16H20Cl2N8O2S, molecular weight 459.35 g/mol.
  • Available in small pack sizes suitable for research (for example, 5 mg).
  • Soluble in DMSO; supplied as the dihydrochloride salt.
  • Used in myocardial infarction and platelet aggregation research.

Catalog No. 50-002-34329 Supplier Medchemexpress LLC Supplier No. HY119350B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zalunfiban dihydrochloride is a potent, selective platelet αIIbβ3 antagonist used in research applications such as myocardial infarction studies. It is supplied as the dihydrochloride salt with high reported purity and is typically used in biochemical and pharmacological assays.

  • Potent αIIbβ3 antagonist (IC50 = 45 nM).
  • Reported purity 99.07%.
  • Molecular formula C16H20Cl2N8O2S, molecular weight 459.35 g/mol.
  • Available in small pack sizes suitable for research (for example, 5 mg).
  • Soluble in DMSO; supplied as the dihydrochloride salt.
  • Used in myocardial infarction and platelet aggregation research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.