Learn More
Medchemexpress LLC Zalunfiban dihydrochloride | 2815778-41-5 | 99.1% | 50 MG
SDP
Catalog No. 5000234026
Encompass_Preferred
Zalunfiban dihydrochloride is the dihydrochloride salt of zalunfiban (RUC-4), a selective small-molecule antagonist of platelet αIIbβ3 used in research on platelet inhibition and myocardial infarction. The compound shows reported potency with IC50 ≈ 45 nM and is supplied as a high-purity research reagent.
  • Selective platelet αIIbβ3 antagonist (IC50 ≈ 45 nM)
  • Suitable for preclinical and in vitro myocardial infarction research
  • Provided as a dihydrochloride salt for improved stability and solubility
  • High purity (reported ~99.1%)
  • Soluble in DMSO for experimental use
Catalog No. 50-002-34026 Supplier Medchemexpress LLC Supplier No. HY119350B50MG
Zalunfiban dihydrochloride is the dihydrochloride salt of zalunfiban (RUC-4), a selective small-molecule antagonist of platelet αIIbβ3 used in research on platelet inhibition and myocardial infarction. The compound shows reported potency with IC50 ≈ 45 nM and is supplied as a high-purity research reagent.
  • Selective platelet αIIbβ3 antagonist (IC50 ≈ 45 nM)
  • Suitable for preclinical and in vitro myocardial infarction research
  • Provided as a dihydrochloride salt for improved stability and solubility
  • High purity (reported ~99.1%)
  • Soluble in DMSO for experimental use