Learn More
Newcomer Supply Zamboni Fixative, 1 Liter. For Light & Electron Microscopy
SDP
Catalog No. NC9335034
Encompass
Newcomer Supply Zamboni Fixative is a ready-to-use phosphate buffered picric acid-formaldehyde (PAF) fixative with applications for light and electron microscopy.  This solution is very stable and provides good general fixation with rapid penetration and optimal preservation and stabilization of cellular proteins. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC9335034 Supplier Newcomer Supply Supplier No. 1459A
Newcomer Supply Zamboni Fixative is a ready-to-use phosphate buffered picric acid-formaldehyde (PAF) fixative with applications for light and electron microscopy.  This solution is very stable and provides good general fixation with rapid penetration and optimal preservation and stabilization of cellular proteins. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.