Learn More
StatLab Zamboni's Fixative

Description
- Zambonis Fixative stabilizes cellular proteins, is not easily destroyed by tissue fluids, and will not cause tissue deterioration
- This allows specimens to be kept at room temperature indefinitely and be sent through the mail without the surgical samples being compromised
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Chemical Solution |
| Product Type | Fixative |
| Quantity | 1000 mL |
| Solution Type | Fixative |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.