Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Zamboni's Fixative
SDP

Catalog No. 21033375
Change view
Click to view available options
Quantity:
16 oz.
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-375 16 oz.
21-033-374 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-375 Supplier StatLab Supplier No. FXZAMPT
Add to Cart
Add to Cart

Excellent for ER or PR immunostains and electron microscopy

  • Zambonis Fixative stabilizes cellular proteins, is not easily destroyed by tissue fluids, and will not cause tissue deterioration
  • This allows specimens to be kept at room temperature indefinitely and be sent through the mail without the surgical samples being compromised

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 16 oz.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.