Learn More
Medchemexpress LLC HY-107649 10mg Medchemexpress, Zamifenacin (fumarate) CAS:127308-98-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10764910MG
Encompass_Preferred
Medchemexpress, HY-107649 10mg Zamifenacin (fumarate) CAS:127308-98-9 Zamifenacin fumarate (UK-76654 fumarate) is a potent gut-selective muscarinic M3 receptor antagonist. Zamifenacin significantly reduces colonic motility in irritable bowel syndrome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9605