Learn More
Medchemexpress LLC Zamifenacin fumarate | 127308-98-9 | 99.0% | 531.60 g·mol^-1 | C31H33NO7 | 50 MG
SDP
Catalog No. 5000240310
Encompass_Preferred
Zamifenacin fumarate is a gut-selective muscarinic M3 receptor antagonist used in research to reduce colonic motility. Supplied as a high-purity solid, it is intended for laboratory pharmacology and GPCR signaling studies.
  • Gut-selective muscarinic M3 receptor antagonist for research use.
  • Shown to reduce colonic motility in preclinical studies.
  • High reported purity (99.0%).
  • Solid, white to off-white appearance suitable for analytical handling.
  • Available in multiple pack sizes, including 50 MG.
  • Store sealed and away from moisture; recommended refrigerated storage and defined solution storage temperatures.
Catalog No. 50-002-40310 Supplier Medchemexpress LLC Supplier No. HY10764950MG
Zamifenacin fumarate is a gut-selective muscarinic M3 receptor antagonist used in research to reduce colonic motility. Supplied as a high-purity solid, it is intended for laboratory pharmacology and GPCR signaling studies.
  • Gut-selective muscarinic M3 receptor antagonist for research use.
  • Shown to reduce colonic motility in preclinical studies.
  • High reported purity (99.0%).
  • Solid, white to off-white appearance suitable for analytical handling.
  • Available in multiple pack sizes, including 50 MG.
  • Store sealed and away from moisture; recommended refrigerated storage and defined solution storage temperatures.