Learn More
Medchemexpress LLC Zamifenacin fumarate | 127308-98-9 | 99.0% | 531.60 g/mol | C31H33NO7 | 1 ML
SDP
Catalog No. 5000240104
Encompass_Preferred
Zamifenacin fumarate is a gut-selective muscarinic M3 receptor antagonist provided for research use. It reduces colonic motility and is commonly used in pharmacology studies of muscarinic receptor function. The compound is available as a solid or as pre-made DMSO solution (typical 10 mM, 1 mL), with reported purity around 99.0% and molecular weight 531.60 g/mol.
Catalog No. 50-002-40104 Supplier Medchemexpress LLC Supplier No. HY10764910MM 1ML
Zamifenacin fumarate is a gut-selective muscarinic M3 receptor antagonist provided for research use. It reduces colonic motility and is commonly used in pharmacology studies of muscarinic receptor function. The compound is available as a solid or as pre-made DMSO solution (typical 10 mM, 1 mL), with reported purity around 99.0% and molecular weight 531.60 g/mol.