Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zamifenacin fumarate | 127308-98-9 | 99.0% | 531.60 g/mol | C31H33NO7 | 25 MG
SDP

Catalog No. 5000240309
Encompass_Preferred

Zamifenacin fumarate is the fumarate salt of zamifenacin, a gut-selective muscarinic M3 receptor antagonist used in research to study gastrointestinal motility. Supplied as a solid for laboratory research use only; not for human or clinical use.

  • Potent gut-selective muscarinic M3 receptor antagonist.
  • High purity: 99.0% (HPLC).
  • White to off-white solid form for easy handling.
  • Molecular weight 531.60 g/mol.
  • Soluble in DMSO (100 mg/mL) and sparingly soluble in water with heating or ultrasonic treatment.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (1 month).

Catalog No. 50-002-40309 Supplier Medchemexpress LLC Supplier No. HY10764925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zamifenacin fumarate is the fumarate salt of zamifenacin, a gut-selective muscarinic M3 receptor antagonist used in research to study gastrointestinal motility. Supplied as a solid for laboratory research use only; not for human or clinical use.

  • Potent gut-selective muscarinic M3 receptor antagonist.
  • High purity: 99.0% (HPLC).
  • White to off-white solid form for easy handling.
  • Molecular weight 531.60 g/mol.
  • Soluble in DMSO (100 mg/mL) and sparingly soluble in water with heating or ultrasonic treatment.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.