Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zamifenacin fumarate | 127308-98-9 | 99.0% | 531.60 g/mol | C31H33NO7 | 5 MG
SDP

Catalog No. 5000240321
Encompass_Preferred

Zamifenacin fumarate is the fumarate salt of zamifenacin (UK-76654), a gut-selective muscarinic M3 receptor antagonist used in research to study gastrointestinal motility. It is provided as a high-purity solid for use in in vitro and in vivo pharmacology studies targeting colonic motility.

  • Potent gut-selective muscarinic M3 receptor antagonist that reduces colonic motility.
  • High purity suitable for research applications (99.0%).
  • Solid, white to off-white form for convenient handling and dosing.
  • Molecular weight 531.60 g/mol, molecular formula C31H33NO7.
  • Recommended storage at 4°C; in solution, -80°C for long-term stability.

Catalog No. 50-002-40321 Supplier Medchemexpress LLC Supplier No. HY1076495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zamifenacin fumarate is the fumarate salt of zamifenacin (UK-76654), a gut-selective muscarinic M3 receptor antagonist used in research to study gastrointestinal motility. It is provided as a high-purity solid for use in in vitro and in vivo pharmacology studies targeting colonic motility.

  • Potent gut-selective muscarinic M3 receptor antagonist that reduces colonic motility.
  • High purity suitable for research applications (99.0%).
  • Solid, white to off-white form for convenient handling and dosing.
  • Molecular weight 531.60 g/mol, molecular formula C31H33NO7.
  • Recommended storage at 4°C; in solution, -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.