Learn More
Medchemexpress LLC Zamifenacin fumarate | 127308-98-9 | MFCD09263617 | 99.0% | 531.60 g/mol | C31H33NO7 | 100 MG
SDP
Catalog No. 5000240326
Encompass_Preferred
Zamifenacin fumarate is a synthetic small molecule and gut-selective muscarinic M3 receptor antagonist used in pharmacological research to study gastrointestinal motility. It has been shown to reduce colonic motility in preclinical studies. The fumarate salt has formula C31H33NO7, molecular weight 531.60 g/mol, CAS 127308-98-9, and is supplied at high purity for research applications.
  • Selective muscarinic M3 receptor antagonism.
  • Gut-selective activity that minimizes systemic muscarinic effects.
  • Demonstrated reduction of colonic motility in preclinical models.
  • Supplied as the fumarate salt with molecular formula C31H33NO7.
  • High purity suitable for research use (99.0%).
Catalog No. 50-002-40326 Supplier Medchemexpress LLC Supplier No. HY107649100MG
Zamifenacin fumarate is a synthetic small molecule and gut-selective muscarinic M3 receptor antagonist used in pharmacological research to study gastrointestinal motility. It has been shown to reduce colonic motility in preclinical studies. The fumarate salt has formula C31H33NO7, molecular weight 531.60 g/mol, CAS 127308-98-9, and is supplied at high purity for research applications.
  • Selective muscarinic M3 receptor antagonism.
  • Gut-selective activity that minimizes systemic muscarinic effects.
  • Demonstrated reduction of colonic motility in preclinical models.
  • Supplied as the fumarate salt with molecular formula C31H33NO7.
  • High purity suitable for research use (99.0%).