Learn More
Abcam Zamifenacin fumarate, M3 receptor antagonist, 10MG
Catalog No. 502706543
Encompass_Preferred
MW 531.6 g/mol, Purity >99%. Selective M₃ receptor antagonist (pKi values are 8.52 (M₃), 7.93 (M₂), 7.90 (M₁) and 7.78 (M₄) respectively). Inhibits gastrointestinal motility in vivo without causing cardiovascular effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6543 Supplier Abcam Supplier No. AB14151110MG
MW 531.6 g/mol, Purity >99%. Selective M₃ receptor antagonist (pKi values are 8.52 (M₃), 7.93 (M₂), 7.90 (M₁) and 7.78 (M₄) respectively). Inhibits gastrointestinal motility in vivo without causing cardiovascular effects.
The product is subject to the following: Abcam Restricted Use Statement