Learn More
Cayman Chemical Zamifenacin (fumarate), Purity: ≥98%, CAS: 127308-98-9, 10 mg
SDP
Catalog No. 502950872
Encompass_Preferred
An antagonist of M3 muscarinic acetylcholine receptors (Ki = 10 nM for the human receptor); selective for M3 over M1, M2, M4, and M5 receptors in radioligand binding assays (Kis = 55, 153, 68, and 34 nM, respectively); inhibits cholecystokinin-induced small bowel motility (ED50 = 0.9 mg/kg) but does not increase heart rate (ED150 = 80.0 mg/kg) in dogs
Catalog No. 50-295-0872 Supplier Cayman Chemical Supplier No. 3284810 MG
An antagonist of M3 muscarinic acetylcholine receptors (Ki = 10 nM for the human receptor); selective for M3 over M1, M2, M4, and M5 receptors in radioligand binding assays (Kis = 55, 153, 68, and 34 nM, respectively); inhibits cholecystokinin-induced small bowel motility (ED50 = 0.9 mg/kg) but does not increase heart rate (ED150 = 80.0 mg/kg) in dogs