Learn More
Medchemexpress LLC HY-13210 10mg Medchemexpress, Zanamivir CAS:139110-80-8 Purity:>98%
SDP
Catalog No. 502081306
Encompass_Preferred
Medchemexpress, HY-13210 10mg Zanamivir CAS:139110-80-8 Zanamivir is an influenza viral neuraminidase inhibitor with IC50 values of 0.95 nM and 2.7 nM for influenza A and B, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1306 Supplier Medchemexpress LLC Supplier No. HY1321010MG
Medchemexpress, HY-13210 10mg Zanamivir CAS:139110-80-8 Zanamivir is an influenza viral neuraminidase inhibitor with IC50 values of 0.95 nM and 2.7 nM for influenza A and B, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.