Learn More
Apexbio Technology LLC Zanamivir, 139110-80-8, MFCD00866966, 1 g
SDP
Supplier:  Apexbio Technology LLC B21361000
Encompass_Preferred
MF: C12H20N4O7, MW: 332.31, Solubility: insoluble in DMSO, 8.99 mg/mL in H2O. Zanamivir is a selective inhibitor of influenza A and B virus neuraminidases with IC50 values of 0.64-7.9nM.
Catalog No. 50-101-1552