Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zanamivir monohydrate

Catalog No. AC462960010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462960010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462960010 Supplier Thermo Scientific Chemicals Supplier No. 462960010
  • $490.32
    $416.65 / Each of 1

    Save $73.67

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1260601-68-0 | C12 H20 N4 O7 . H2 O | 350.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1260601-68-0
Molecular Formula C12 H20 N4 O7 . H2 O
Molecular Weight (g/mol) 350.33
MDL Number MFCD00866966

Specifications

Alpha Vector ZANAMIVIR
Melting Point 256°C
Color White
Loss on Drying 4 % to 9 % (1 g, 105°C) (vacuum)
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
Specific Rotation 36° to 38° (20°C, 589 nm) (1 % aq. soln.)(20°C, 589 nm) (on dry substance)
Formula Weight 350.33
Physical Form Crystalline Powder
Chemical Name or Material Zanamivir monohydrate
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 1 year
storageNote1 Refrigerator +4°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.