Learn More
TARGETMOL CHEMICALS INC ZANDATRIGINE 5MG
SDP
Catalog No. 502580044
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zandatrigine (NBI-921352) is a blocker of sodium channel protein type 8 subunit alpha and can be used in studies about nervous system pathologies of epilepsy treatment. purity: 99%
Catalog No. 50-258-0044 Supplier TARGETMOL CHEMICALS INC Supplier No. T396305MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zandatrigine (NBI-921352) is a blocker of sodium channel protein type 8 subunit alpha and can be used in studies about nervous system pathologies of epilepsy treatment. purity: 99%