Learn More
Medchemexpress LLC Zanubrutinib | 1691249-45-2 | MFCD31567461 | 471.55 | C27H29N5O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101474A25MG
Encompass_Preferred
Zanubrutinib is a selective, irreversible small-molecule inhibitor of Bruton tyrosine kinase (BTK) used in research to study BTK signaling and pharmacology. The compound shows high biochemical potency and is supplied as a solid reagent suitable for analytical, biochemical, and cellular studies. This catalog item is provided for research use only.
Catalog No. 50-002-47928