Learn More
TARGETMOL CHEMICALS INC ZANUBRUTINIB 5MG
Supplier: TARGETMOL CHEMICALS INC TQ00395MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (±)-Zanubrutinib ((±)-BGB-3111) is a potent and orally available Bruton's tyrosine kinase (Btk) inhibitor that demonstrates superior oral bioavailability achieving higher exposure and more complete target inhibi. purity: 98%