Learn More
TARGETMOL CHEMICALS INC ZANUBRUTINIB 5MG
SDP
Catalog No. 502579178
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zanubrutinib (BGB-3111) is an inhibitor of Bruton tyrosine kinase (BTK). purity: 99%
Catalog No. 50-257-9178 Supplier TARGETMOL CHEMICALS INC Supplier No. T75845MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zanubrutinib (BGB-3111) is an inhibitor of Bruton tyrosine kinase (BTK). purity: 99%