Learn More
Medchemexpress LLC HY-101474A 5mg Medchemexpress, Zanubrutinib CAS:1691249-45-2 Purity:>98%
SDP
Catalog No. 501967851
Encompass_Preferred
Medchemexpress, HY-101474A 5mg Zanubrutinib CAS:1691249-45-2 Zanubrutinib (BGB-3111) is a selective Bruton tyrosine kinase (BTK) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7851 Supplier Medchemexpress LLC Supplier No. HY101474A5MG
Medchemexpress, HY-101474A 5mg Zanubrutinib CAS:1691249-45-2 Zanubrutinib (BGB-3111) is a selective Bruton tyrosine kinase (BTK) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.