Learn More
Selleck Chemical LLC Zanubrutinib (BGB-3111) 2mg 1691249-45-2
SDP
Supplier:  Selleck Chemical LLC S87912MG
Encompass_Preferred
Zanubrutinib (BGB-3111) is a potent, specific and irreversible BTK inhibitor that has been shown to have a lower off-target inhibitory activity on other kinases, including ITK, JAK3 and EGFR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8310