Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zanubrutinib-d5 | 00-00-0 | 99.0% | 476.58 g/mol | C27H24D5N5O3 | 1 MG
SDP

Catalog No. 5000249185
Encompass_Preferred

Zanubrutinib-d5 is a deuterium-labeled analogue of zanubrutinib intended for research use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, and LC-MS. It is supplied as a solid for laboratory studies and therapeutic drug monitoring method development.

  • Deuterium-labeled internal standard for quantitative analysis
  • Purity: 99.0%
  • Molecular formula: C27H24D5N5O3; molecular weight: 476.58 g/mol
  • Appearance: white to off-white solid
  • Solubility: DMSO 30 mg/mL; may require ultrasonic and warming
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Packaged in small-mass units suitable for analytical standards (1 mg available)

Catalog No. 50-002-49185 Supplier Medchemexpress LLC Supplier No. HY101474S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zanubrutinib-d5 is a deuterium-labeled analogue of zanubrutinib intended for research use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, and LC-MS. It is supplied as a solid for laboratory studies and therapeutic drug monitoring method development.

  • Deuterium-labeled internal standard for quantitative analysis
  • Purity: 99.0%
  • Molecular formula: C27H24D5N5O3; molecular weight: 476.58 g/mol
  • Appearance: white to off-white solid
  • Solubility: DMSO 30 mg/mL; may require ultrasonic and warming
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Packaged in small-mass units suitable for analytical standards (1 mg available)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.