Learn More
Medchemexpress LLC Zanzalintinib | 2367004-54-2 | C29H25FN4O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13869610MG
Encompass_Preferred
Zanzalintinib is an orally active, ATP-competitive multi-receptor tyrosine kinase inhibitor used for preclinical research. It targets MET, VEGFR2, AXL, and MER, with reported cell-based IC50s in the low nanomolar range, and is provided as a research-grade compound (molecular formula C29H25FN4O5; MW 528.53). This compound is for laboratory research only and is not intended for human use.
  • Multi-kinase activity against MET, VEGFR2, AXL, and MER.
  • Potent cellular activity with low-nanomolar IC50 values.
  • Suitable for kinase inhibition and cell-based assays.
  • High purity for research applications (>98% reported).
  • Stable under recommended storage when protected from light.
  • Provided as a solid for preparation of stock solutions.
Catalog No. 50-002-60044