Learn More
Medchemexpress LLC Zanzalintinib (XL092) | 2367004-54-2 | MFCD34179545 | 99.9% | 528.53 g/mol | C29H25FN4O5 | 5 MG
SDP
Catalog No. 5000221158
Encompass_Preferred
Zanzalintinib (XL092) is an orally active, ATP-competitive inhibitor of multiple receptor tyrosine kinases, supplied as a solid research-grade compound for in vitro and preclinical studies. Not for human or clinical use.
  • Orally active, ATP-competitive multi-kinase inhibitor.
  • Potent activity against MET, VEGFR2, AXL, and MER (low-nanomolar range).
  • High purity (≈99.9%) suitable for biochemical and cellular assays.
  • Available in small pack sizes for assay development and screening.
  • Solid form, stable for storage and experimental workflows.
Catalog No. 50-002-21158 Supplier Medchemexpress LLC Supplier No. HY1386965MG
Zanzalintinib (XL092) is an orally active, ATP-competitive inhibitor of multiple receptor tyrosine kinases, supplied as a solid research-grade compound for in vitro and preclinical studies. Not for human or clinical use.
  • Orally active, ATP-competitive multi-kinase inhibitor.
  • Potent activity against MET, VEGFR2, AXL, and MER (low-nanomolar range).
  • High purity (≈99.9%) suitable for biochemical and cellular assays.
  • Available in small pack sizes for assay development and screening.
  • Solid form, stable for storage and experimental workflows.