Learn More
Eurofins Discoverx ZAP-70, active, 20 µg. (14-404)
Supplier:  Eurofins Discoverx 14404
Encompass
C-terminal 6xHis-tagged recombinant, human full length ZAP-70. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8789