Learn More
CAPRICO BIOTECHNOLOGIES INC ZAP-70 iFluor488 Antibody Conjugate
Catalog No. 502279300
Encompass_Preferred
The ZAP-70 (IFluor488) Antibody Conjugate from Caprico Biotechnologies is an antibody that targets ZAP-70, a tyrosine kinase that plays a key role in T cell receptor signaling. This antibody is conjugated to IFluor488, a green fluorescent dye, allowing for direct detection of ZAP-70 expression via immunofluorescence. The antibody is made in rabbit and is supplied as a liquid in PBS with 0.1% sodium azide and 50% glycerol. The immunogen is a synthetic peptide from human ZAP-70. This antibody can be used for a variety of applications including Western blot, immunoprecipitation, immunofluorescence, and flow cytometry. It has been tested for use in human, mouse, and rat samples.
This item is not returnable. View return policy
Catalog No. 50-227-9300 Supplier CAPRICO BIOTECHNOLOGIES INC Supplier No. 4203115
The ZAP-70 (IFluor488) Antibody Conjugate from Caprico Biotechnologies is an antibody that targets ZAP-70, a tyrosine kinase that plays a key role in T cell receptor signaling. This antibody is conjugated to IFluor488, a green fluorescent dye, allowing for direct detection of ZAP-70 expression via immunofluorescence. The antibody is made in rabbit and is supplied as a liquid in PBS with 0.1% sodium azide and 50% glycerol. The immunogen is a synthetic peptide from human ZAP-70. This antibody can be used for a variety of applications including Western blot, immunoprecipitation, immunofluorescence, and flow cytometry. It has been tested for use in human, mouse, and rat samples.