Learn More
Biocare Medical, LLC ZAP-70 Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC ACI259A
Encompass_Preferred
Zeta-associated protein-70 (ZAP-70 antibody) is a tyrosine kinase normally expressed by natural killer cells and T cells. Several studies have indicated a correlation between ZAP-70 expression and immunoglobulin heavy-chain variable-region (IgVH) mutational status in the leukemic cells of chronic lymphocytic leukemia (CLL), with ZAP-70 suggested as a surrogate marker for IgHV mutational status. The mutational status of IgVH genes in CLL is an important prognostic factor in the disease and ZAP-70 overexpression indicates an unfavorable disease course in terms of progression and overall survival.
Catalog No. 50-826-25