Learn More
Enzo Life Sciences Zap 70 monoclonal antibody (11H40) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7090100
Encompass_Preferred
Clone: 11H40. Isotype: IgG1κ. Immunogen: Recombinant full length human Zap-70. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Rat. Applications: IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3247