Learn More
Carnabio Usa Inc ZAP70/10UG/FULL-LENGTH
Catalog No. 507541033
Encompass
ZAP70, Full-length, Wild type, Amino Acid 1-619, NP_001070.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1033 Supplier Carnabio Usa Inc Supplier No. 0817710
ZAP70, Full-length, Wild type, Amino Acid 1-619, NP_001070.2, Expressed in Insect (sf21); 10microg