Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7535MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: ZAP70/528. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Jurkat cells. Tonsil or lymph node. Cellular Staining: Cytoplasmic. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human ZAP70 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 70kDa.Chromosome: 2q12. Gene Symbol: ZAP70. Gene ID: 7535. Ensemble: ENSG00000115085. Uniprot: NP_001070.
Catalog No. 50-188-5293