Learn More
Aviva Systems Biology Corp ZAP70 Antibody - N-terminal region (ARP61186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZAP70. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZAP70. Alternate names: FLJ17670, FLJ17679, SRK, STD, TZK, ZAP-70ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-287-289