Learn More
Medchemexpress LLC Zapa sulfate | 371962-01-5 | >98.0% | 244.25 g/mol | C4H8N2O6S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100799100MG
Encompass_Preferred
ZAPA sulfate is the sulfate salt of a small-molecule agonist at low-affinity GABAA receptors used for laboratory research. It induces membrane hyperpolarization in Ascaris muscle cells with a reported EC50 of 10.3 μM and is supplied as a solid and as a 10 mM solution in DMSO for convenient experimental use.
  • Agonist at low-affinity GABAA receptors.
  • Reported EC50 of 10.3 μM for Ascaris muscle cell hyperpolarization.
  • Available as solid and 10 mM solution in DMSO.
  • Molecular weight 244.25 g/mol.
  • Datasheet and safety data sheet available for handling information.
Catalog No. 50-002-13267