Learn More
Medchemexpress LLC Zapa sulfate | 371962-01-5 | 99.3% | 244.25 g·mol-1 | C4H8N2O6S2 | 1 ML

Supplier: Medchemexpress LLC HY10079910MM 1ML
ZAPA sulfate is a small-molecule agonist of low-affinity GABAA receptors that induces membrane hyperpolarization in Ascaris muscle cells (reported EC50 = 10.3 μM). It is supplied as a 10 mM solution in DMSO for in vitro research applications.
- Agonist at low-affinity GABAA receptors.
- Induces membrane hyperpolarization in Ascaris muscle cells (EC50 = 10.3 μM).
- Supplied as a 10 mM solution in DMSO for immediate use.
- Packaged as a 1 mL aliquot suitable for assay preparation.
- High purity confirmed by LCMS (99.25%).
- Molecular weight 244.25 g·mol-1; formula C4H8N2O6S2.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.