Learn More
Medchemexpress LLC Zapa sulfate | 371962-01-5 | 99.3% | 244.25 g/mol | C4H8N2O6S2 | 10 MG
SDP
Catalog No. 5000212196
Encompass_Preferred
Zapa sulfate is the sulfate salt of ZAPA, a research compound that acts as an agonist at low-affinity GABAA receptors. It induces membrane hyperpolarization in Ascaris muscle cells with a reported EC50 of 10.3 μM and is provided as a solid sulfate salt for research applications.
  • Acts as an agonist at low-affinity GABAA receptors.
  • Induces membrane hyperpolarization in Ascaris muscle cells (EC50 10.3 μM).
  • High reported purity (99.25%).
  • Molecular weight 244.25 g/mol; formula C4H8N2O6S2.
  • Supplied as a solid sulfate salt suitable for in vitro research.
Catalog No. 50-002-12196 Supplier Medchemexpress LLC Supplier No. HY10079910MG
Zapa sulfate is the sulfate salt of ZAPA, a research compound that acts as an agonist at low-affinity GABAA receptors. It induces membrane hyperpolarization in Ascaris muscle cells with a reported EC50 of 10.3 μM and is provided as a solid sulfate salt for research applications.
  • Acts as an agonist at low-affinity GABAA receptors.
  • Induces membrane hyperpolarization in Ascaris muscle cells (EC50 10.3 μM).
  • High reported purity (99.25%).
  • Molecular weight 244.25 g/mol; formula C4H8N2O6S2.
  • Supplied as a solid sulfate salt suitable for in vitro research.