Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zapa sulfate | 371962-01-5 | 99.3% | 244.25 g/mol | C4H8N2O6S2 | 25 MG
SDP

Catalog No. 5000212543
Encompass_Preferred

ZAPA sulfate is a research-grade sulfate salt that functions as an agonist at low-affinity GABAA receptors and induces membrane hyperpolarization in Ascaris muscle cells. It is provided as a solid for use in in vitro pharmacology and electrophysiology studies.

  • High purity: 99.3%.
  • Sulfate salt formulation.
  • Molecular weight 244.25 g/mol.
  • Chemical formula C4H8N2O6S2.
  • CAS number 371962-01-5.
  • Reported EC50 ≈ 10.3 μM for Ascaris muscle cell hyperpolarization.
  • Supplied in solid pack sizes including 25 MG.
  • Suitable for in vitro pharmacology and electrophysiology applications.

Catalog No. 50-002-12543 Supplier Medchemexpress LLC Supplier No. HY10079925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZAPA sulfate is a research-grade sulfate salt that functions as an agonist at low-affinity GABAA receptors and induces membrane hyperpolarization in Ascaris muscle cells. It is provided as a solid for use in in vitro pharmacology and electrophysiology studies.

  • High purity: 99.3%.
  • Sulfate salt formulation.
  • Molecular weight 244.25 g/mol.
  • Chemical formula C4H8N2O6S2.
  • CAS number 371962-01-5.
  • Reported EC50 ≈ 10.3 μM for Ascaris muscle cell hyperpolarization.
  • Supplied in solid pack sizes including 25 MG.
  • Suitable for in vitro pharmacology and electrophysiology applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.