Learn More
Medchemexpress LLC Zapa (sulfate) | 371962-01-5 | 99.3% | 244.25 g/mol | C4H8N2O6S2 | 5 MG
SDP
Catalog No. 5000213426
Encompass_Preferred
Zapa (sulfate) is a small-molecule agonist of low-affinity GABAA receptors used as a research reagent to provoke membrane hyperpolarization in biological models. Reported activity includes induction of hyperpolarization in Ascaris muscle cells with an EC50 of 10.3 μM. The compound is supplied as a purified solid for laboratory pharmacology and analytical studies.
  • Agonist at low-affinity GABAA receptors.
  • Induces membrane hyperpolarization in Ascaris muscle cells (EC50 10.3 μM).
  • High purity (~99.25%) suitable for analytical use.
  • Offered in small, research-scale quantities for assay development.
  • Molecular weight 244.25 g/mol; molecular formula C4H8N2O6S2.
Catalog No. 50-002-13426 Supplier Medchemexpress LLC Supplier No. HY1007995MG
Zapa (sulfate) is a small-molecule agonist of low-affinity GABAA receptors used as a research reagent to provoke membrane hyperpolarization in biological models. Reported activity includes induction of hyperpolarization in Ascaris muscle cells with an EC50 of 10.3 μM. The compound is supplied as a purified solid for laboratory pharmacology and analytical studies.
  • Agonist at low-affinity GABAA receptors.
  • Induces membrane hyperpolarization in Ascaris muscle cells (EC50 10.3 μM).
  • High purity (~99.25%) suitable for analytical use.
  • Offered in small, research-scale quantities for assay development.
  • Molecular weight 244.25 g/mol; molecular formula C4H8N2O6S2.