Learn More
Medchemexpress LLC Zapa (sulfate) | 371962-01-5 | 99.3% | 244.25 g/mol | C4H8N2O6S2 | 50 MG
SDP
Catalog No. 5000211184
Encompass_Preferred
ZAPA sulfate is a research reagent used to probe GABAergic signaling; it acts as an agonist at low-affinity GABAA receptors and induces membrane hyperpolarization in Ascaris muscle cells with a reported EC50 of ~10.3 μM.
  • Agonist at low-affinity GABAA receptors.
  • Induces Ascaris muscle cell membrane hyperpolarization; EC50 ≈ 10.3 μM.
  • High supplied purity (~99.25%).
  • Molecular weight 244.25 g/mol.
  • Available in small research quantities, including 50 MG and 10 mM solution formats.
Catalog No. 50-002-11184 Supplier Medchemexpress LLC Supplier No. HY10079950MG
ZAPA sulfate is a research reagent used to probe GABAergic signaling; it acts as an agonist at low-affinity GABAA receptors and induces membrane hyperpolarization in Ascaris muscle cells with a reported EC50 of ~10.3 μM.
  • Agonist at low-affinity GABAA receptors.
  • Induces Ascaris muscle cell membrane hyperpolarization; EC50 ≈ 10.3 μM.
  • High supplied purity (~99.25%).
  • Molecular weight 244.25 g/mol.
  • Available in small research quantities, including 50 MG and 10 mM solution formats.