Learn More
Andwin Scientific ZAPCAP CR PTFE .45 12/CS
SDP
Catalog No. 50465467
Encompass_Preferred
The Whatman ZapCap bottle-top filter is an efficient laboratory supply for filtering HPLC solutions. It is specially designed to have a great resistance against chemicals, even agressive samples. This product is sold with a borosilicate prefilter that can be utilized for high flow rates. Its connections are compatible with any standard bottle with a 33mm or 45mm neck size. This bottle-top filter has a diameter of 76 mm and filter area of 39.2 cm.
Catalog No. 50-465-467 Supplier Andwin Scientific Supplier No. 74375
The Whatman ZapCap bottle-top filter is an efficient laboratory supply for filtering HPLC solutions. It is specially designed to have a great resistance against chemicals, even agressive samples. This product is sold with a borosilicate prefilter that can be utilized for high flow rates. Its connections are compatible with any standard bottle with a 33mm or 45mm neck size. This bottle-top filter has a diameter of 76 mm and filter area of 39.2 cm.