Learn More
Cayman Chemical Zapnometinib, Purity: ≥95%, CAS: 303175-44-2, 10 mg
SDP
Catalog No. 502916726
Encompass_Preferred
A MEK inhibitor (IC50 = 5.73 nM) and an active metabolite of CI-1040; reduces viral titers in A549 and MDCK cells infected with influenza A (EC50s = 6.356 and 4.498 µM, respectively); increases survival in H1N1pdm09 influenza A-infected mice at 50 mg/kg per day
Catalog No. 50-291-6726 Supplier Cayman Chemical Supplier No. 3851710 MG
A MEK inhibitor (IC50 = 5.73 nM) and an active metabolite of CI-1040; reduces viral titers in A549 and MDCK cells infected with influenza A (EC50s = 6.356 and 4.498 µM, respectively); increases survival in H1N1pdm09 influenza A-infected mice at 50 mg/kg per day